siRNA Details
Gene Symbol: A130023I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037526.1 | - | 1 | 1096 | ||
AK088084.1 | - | 2521 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A130023I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037526.1 | - | 1 | 1096 | ||
AK088084.1 | - | 2521 |