siRNA Details
| Gene Symbol: 4930412M03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029606.1 | - | 1, 2 | 519 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177098.1 | - | 519 | Chr. - - [-] on Build | ||

4930412M03Rik
Genomic Map