siRNA Details
| Gene Symbol: 4930412M03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029606.1 | - | 1 | 498 | ||
| AK089395.1 | - | 1 | 515 | |||
| AK154788.1 | - | 1 | 515 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177098.1 | - | 498 | Chr. - - [-] on Build | ||

4930412M03Rik
Genomic Map