siRNA Details
Gene Symbol: Gm5087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121589.1 | 5 | 583 | |||
GenBank mRNA | AK081144.1 | - | 5 | 576 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121589.1 | 5 | 583 | |||
GenBank mRNA | AK081144.1 | - | 5 | 576 |