siRNA Details
| Gene Symbol: Gm14964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045662.1 | - | 1 | 1357 | ||
| AK045826.1 | - | 2 | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14964| Gene Symbol: Gm14964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045662.1 | - | 1 | 1357 | ||
| AK045826.1 | - | 2 | 399 | |||