siRNA Details
| Gene Symbol: 8030423F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045738.1 | 3 | 914 | |||
| GenBank mRNA | AK033101.1 | - | 3 | 915 | ||
| AK078739.1 | - | 1 | 585 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8030423F21Rik| Gene Symbol: 8030423F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045738.1 | 3 | 914 | |||
| GenBank mRNA | AK033101.1 | - | 3 | 915 | ||
| AK078739.1 | - | 1 | 585 | |||