siRNA Details
| Gene Symbol: E330013P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026942.1 | 3 | 267 | |||
| GenBank mRNA | AK087735.1 | - | 3 | 254 | ||
| AK163106.1 | - | 2 | 341 | |||
| BC076607.1 | - | 3 | 267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E330013P04Rik| Gene Symbol: E330013P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026942.1 | 3 | 267 | |||
| GenBank mRNA | AK087735.1 | - | 3 | 254 | ||
| AK163106.1 | - | 2 | 341 | |||
| BC076607.1 | - | 3 | 267 | |||