siRNA Details
| Gene Symbol: Gm9898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087209.1 | - | 2 | 889 | ||
| BB534251.2 | - | 2 | 401 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9898| Gene Symbol: Gm9898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087209.1 | - | 2 | 889 | ||
| BB534251.2 | - | 2 | 401 | |||