siRNA Details
| Gene Symbol: 6430503K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108091.1 | 3 | 377 | |||
| GenBank mRNA | AK020097.1 | - | 3 | 378 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY718829.1 | - | 3 | 378 | Chr. 2 - 147187424 - 147188407 [+] on Build GRCm38 | |

6430503K07Rik
Genomic Map