siRNA Details
Gene Symbol: Dancr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015531.1 | 2 | 498 | |||
GenBank mRNA | AK045300.1 | - | 739 | |||
AK172090.1 | - | 2 | 498 | |||
AK172703.1 | - | 1 | 681 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dancr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015531.1 | 2 | 498 | |||
GenBank mRNA | AK045300.1 | - | 739 | |||
AK172090.1 | - | 2 | 498 | |||
AK172703.1 | - | 1 | 681 |