siRNA Details
| Gene Symbol: A930005H10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027894.1 | 2 | 367 | |||
| GenBank mRNA | AK137339.1 | - | 1 | 1806 | ||
| BF019418.1 | - | 2 | 367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930005H10Rik| Gene Symbol: A930005H10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027894.1 | 2 | 367 | |||
| GenBank mRNA | AK137339.1 | - | 1 | 1806 | ||
| BF019418.1 | - | 2 | 367 | |||