siRNA Details
| Gene Symbol: Gm9946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049916.1 | - | 3 | 565 | ||
| BB427413.2 | - | 2 | 215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9946| Gene Symbol: Gm9946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049916.1 | - | 3 | 565 | ||
| BB427413.2 | - | 2 | 215 | |||