siRNA Details
| Gene Symbol: 1700041G16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006671.1 | - | 140 | |||
| AK033542.1 | - | 702 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_175196.1 | - | 140 | Chr. - - [-] on Build | ||

1700041G16Rik
Genomic Map