siRNA Details
| Gene Symbol: Gm9962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033504.1 | 3 | 589 | |||
| GenBank mRNA | AK049950.1 | - | 3 | 590 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB653224.1 | - | 3 | 590 | Chr. 7 - 57387272 - 57409168 [+] on Build GRCm38 | |

Gm9962
Genomic Map