siRNA Details
| Gene Symbol: Rbm3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033561.1 | 3 | 604 | |||
| GenBank mRNA | AK013472.1 | - | 3 | 604 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm3os| Gene Symbol: Rbm3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033561.1 | 3 | 604 | |||
| GenBank mRNA | AK013472.1 | - | 3 | 604 | ||