siRNA Details
| Gene Symbol: Hottip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110441.1 | 2 | 1919 | |||
| NR_110442.1 | 2 | 1365 | ||||
| GenBank mRNA | BC046496.1 | - | 2 | 1365 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hottip| Gene Symbol: Hottip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110441.1 | 2 | 1919 | |||
| NR_110442.1 | 2 | 1365 | ||||
| GenBank mRNA | BC046496.1 | - | 2 | 1365 | ||