siRNA Details
| Gene Symbol: Hottip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110441.1 | 2 | 923 | |||
| NR_110442.1 | 2 | 369 | ||||
| GenBank mRNA | BC046496.1 | - | 2 | 369 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF579820.1 | - | 2 | 370 | Chr. 6 - 52262775 - 52266107 [+] on Build GRCm38 | |

Hottip
Genomic Map