siRNA Details
| Gene Symbol: Gata5os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045877.1 | 2 | 238 | |||
| GenBank mRNA | AK018689.1 | - | 2 | 238 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gata5os| Gene Symbol: Gata5os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045877.1 | 2 | 238 | |||
| GenBank mRNA | AK018689.1 | - | 2 | 238 | ||