siRNA Details
Gene Symbol: Gm9999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033461.1 | 4 | 426 | |||
GenBank mRNA | AK076952.1 | - | 3 | 322 | ||
BC050982.1 | - | 196 | ||||
BC100529.1 | - | 1 | 201 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm9999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033461.1 | 4 | 426 | |||
GenBank mRNA | AK076952.1 | - | 3 | 322 | ||
BC050982.1 | - | 196 | ||||
BC100529.1 | - | 1 | 201 |