siRNA Details
| Gene Symbol: Gm9999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033461.1 | 5 | 494 | |||
| GenBank mRNA | AK076952.1 | - | 4 | 390 | ||
| BC050982.1 | - | 264 | ||||
| BC100529.1 | - | 2 | 269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9999| Gene Symbol: Gm9999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033461.1 | 5 | 494 | |||
| GenBank mRNA | AK076952.1 | - | 4 | 390 | ||
| BC050982.1 | - | 264 | ||||
| BC100529.1 | - | 2 | 269 | |||