siRNA Details
| Gene Symbol: Tug1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002321.2 | 4 | 3241 | |||
| NR_002322.2 | 2 | 5229 | ||||
| NR_110490.1 | 3 | 3637 | ||||
| GenBank mRNA | AK088174.1 | - | 4 | 3242 | ||
| BC048828.1 | - | 3 | 3513 | |||
| D50523.1 | - | 1 | 2097 | |||
| DQ517434.1 | - | 2 | 5230 | |||
| DQ517435.1 | - | 4 | 3237 | |||

Tug1
Genomic Map