siRNA Details
| Gene Symbol: Gm11696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038097.1 | 2 | 1784 | |||
| GenBank mRNA | AK028065.1 | - | 2 | 1785 | ||
| BC027814.1 | - | 2 | 1590 | |||
| BC062114.1 | - | 2 | 1747 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11696| Gene Symbol: Gm11696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038097.1 | 2 | 1784 | |||
| GenBank mRNA | AK028065.1 | - | 2 | 1785 | ||
| BC027814.1 | - | 2 | 1590 | |||
| BC062114.1 | - | 2 | 1747 | |||