siRNA Details
| Gene Symbol: A930024E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045820.1 | 3 | 1624 | |||
| GenBank mRNA | AK042918.1 | - | 3 | 1624 | ||
| AK080729.1 | - | 3 | 1542 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930024E05Rik| Gene Symbol: A930024E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045820.1 | 3 | 1624 | |||
| GenBank mRNA | AK042918.1 | - | 3 | 1624 | ||
| AK080729.1 | - | 3 | 1542 | |||