siRNA Details
| Gene Symbol: 9630013D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035877.1 | - | 3 | 1535 | ||
| AK052880.1 | - | 3 | 1561 | |||
| AK080944.1 | - | 1291 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9630013D21Rik| Gene Symbol: 9630013D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035877.1 | - | 3 | 1535 | ||
| AK052880.1 | - | 3 | 1561 | |||
| AK080944.1 | - | 1291 | ||||