siRNA Details
| Gene Symbol: R74862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015529.2 | 1, 2 | 659 | |||
| NR_110354.1 | 1, 2 | 659 | ||||
| NR_110355.1 | 1, 2 | 659 | ||||
| GenBank mRNA | AK034864.1 | - | 1, 2 | 651 | ||
| AK035397.1 | - | 1, 2 | 342 | |||
| AK083267.1 | - | 1, 2 | 391 | |||
| BC005669.1 | - | 1, 2 | 76 | |||
| BC080794.1 | - | 1, 2 | 659 | |||

R74862
Genomic Map