siRNA Details
| Gene Symbol: Gm10069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028592.1 | 3 | 2254 | |||
| GenBank mRNA | AK029950.1 | - | 3 | 1956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10069| Gene Symbol: Gm10069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028592.1 | 3 | 2254 | |||
| GenBank mRNA | AK029950.1 | - | 3 | 1956 | ||