siRNA Details
| Gene Symbol: Fbxl12os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033729.1 | 2 | 429 | |||
| GenBank mRNA | AK017871.1 | - | 2 | 2000 | ||
| AK044040.1 | - | 2 | 424 | |||
| BC087896.1 | - | 2 | 298 | |||
| BC147302.1 | - | 1 | 136 | 9056334 | ||
| BC147303.1 | - | 1 | 136 | 9056351 | ||

Fbxl12os
Genomic Map