siRNA Details
| Gene Symbol: Fbxl12os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033729.1 | 2 | 188 | |||
| GenBank mRNA | AK017871.1 | - | 2 | 1759 | ||
| AK044040.1 | - | 2 | 183 | |||
| BC087896.1 | - | 2 | 57 | |||
| BY123622.1 | - | 2 | 189 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl12os| Gene Symbol: Fbxl12os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033729.1 | 2 | 188 | |||
| GenBank mRNA | AK017871.1 | - | 2 | 1759 | ||
| AK044040.1 | - | 2 | 183 | |||
| BC087896.1 | - | 2 | 57 | |||
| BY123622.1 | - | 2 | 189 | |||