siRNA Details
| Gene Symbol: Dgkeos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110336.1 | 4 | 713 | |||
| GenBank mRNA | AK039819.1 | - | 2 | 620 | ||
| AK044435.1 | - | 678 | ||||
| BB750000.1 | - | 1 | 391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgkeos| Gene Symbol: Dgkeos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110336.1 | 4 | 713 | |||
| GenBank mRNA | AK039819.1 | - | 2 | 620 | ||
| AK044435.1 | - | 678 | ||||
| BB750000.1 | - | 1 | 391 | |||