siRNA Details
| Gene Symbol: Ptpmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012674.1 | - | 4 | 1204 | ||
| AK168416.1 | - | 1080 | ||||
| BC026750.1 | - | 4 | 1193 | 5135841 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpmt1| Gene Symbol: Ptpmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012674.1 | - | 4 | 1204 | ||
| AK168416.1 | - | 1080 | ||||
| BC026750.1 | - | 4 | 1193 | 5135841 | ||