siRNA Details
| Gene Symbol: Gm1976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045963.1 | 5 | 902 | |||
| NR_045964.1 | 4 | 845 | ||||
| GenBank mRNA | AK082664.1 | - | 5 | 902 | ||
| AK137318.1 | - | 4 | 846 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1976| Gene Symbol: Gm1976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045963.1 | 5 | 902 | |||
| NR_045964.1 | 4 | 845 | ||||
| GenBank mRNA | AK082664.1 | - | 5 | 902 | ||
| AK137318.1 | - | 4 | 846 | |||