siRNA Details
Gene Symbol: Gm10248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033550.1 | 1 | 110 | |||
GenBank mRNA | AY512910.1 | - | 1 | 110 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm10248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033550.1 | 1 | 110 | |||
GenBank mRNA | AY512910.1 | - | 1 | 110 |