siRNA Details
| Gene Symbol: Gm11426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033582.1 | 3 | 817 | |||
| GenBank mRNA | AY512914.1 | - | 3 | 817 | ||
| AY512941.1 | - | 3 | 759 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11426| Gene Symbol: Gm11426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033582.1 | 3 | 817 | |||
| GenBank mRNA | AY512914.1 | - | 3 | 817 | ||
| AY512941.1 | - | 3 | 759 | |||