siRNA Details
| Gene Symbol: 4930581F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029475.1 | 2 | 1063 | |||
| GenBank mRNA | AK019824.1 | - | 2 | 1074 | ||
| BC038320.1 | - | 1 | 160 | |||
| CX731663.1 | - | 1 | 338 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930581F22Rik| Gene Symbol: 4930581F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029475.1 | 2 | 1063 | |||
| GenBank mRNA | AK019824.1 | - | 2 | 1074 | ||
| BC038320.1 | - | 1 | 160 | |||
| CX731663.1 | - | 1 | 338 | |||