siRNA Details
| Gene Symbol: 1700086L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030733.1 | 4 | 638 | |||
| NR_030734.1 | 4 | 633 | ||||
| NR_030735.1 | 3 | 411 | ||||
| GenBank mRNA | AK005679.1 | - | 2 | 306 | ||
| AK007020.1 | - | 3 | 407 | |||
| BG083361.2 | - | 4 | 620 | |||

1700086L19Rik
Genomic Map