siRNA Details
| Gene Symbol: 4930417O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045806.1 | 4 | 527 | |||
| GenBank mRNA | AK015160.1 | - | 3 | 539 | ||
| AK076700.1 | - | 4 | 527 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615040.1 | - | 527 | Chr. - - [-] on Build | ||

4930417O22Rik
Genomic Map