siRNA Details
| Gene Symbol: 4930417O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045806.1 | 4 | 995 | |||
| GenBank mRNA | AK015160.1 | - | 3 | 1009 | ||
| AK076700.1 | - | 4 | 995 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930417O22Rik| Gene Symbol: 4930417O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045806.1 | 4 | 995 | |||
| GenBank mRNA | AK015160.1 | - | 3 | 1009 | ||
| AK076700.1 | - | 4 | 995 | |||