siRNA Details
| Gene Symbol: Gm14812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033544.2 | 7 | 1402 | |||
| GenBank mRNA | AK135730.1 | - | 7 | 1404 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14812| Gene Symbol: Gm14812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033544.2 | 7 | 1402 | |||
| GenBank mRNA | AK135730.1 | - | 7 | 1404 | ||