siRNA Details
| Gene Symbol: 9130019P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033635.1 | 11 | 2548 | |||
| GenBank mRNA | AK032212.1 | - | 11 | 2532 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_198118.1 | - | 2532 | Chr. - - [-] on Build | ||
 Back To Top
 Back To Top
 9130019P16Rik
9130019P16Rik Genomic Map
Genomic Map Back To Top
 Back To Top