siRNA Details
| Gene Symbol: Gm10482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136146.1 | - | 1901 | |||
| AK145354.1 | - | 1089 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10482| Gene Symbol: Gm10482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136146.1 | - | 1901 | |||
| AK145354.1 | - | 1089 | ||||