siRNA Details
| Gene Symbol: E330020D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033736.1 | 4 | 1104 | |||
| GenBank mRNA | AK143622.1 | - | 2 | 882 | ||
| AK168480.1 | - | 2 | 880 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E330020D12Rik| Gene Symbol: E330020D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033736.1 | 4 | 1104 | |||
| GenBank mRNA | AK143622.1 | - | 2 | 882 | ||
| AK168480.1 | - | 2 | 880 | |||