siRNA Details
| Gene Symbol: Tmem51os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027137.1 | 1 | 415 | |||
| GenBank mRNA | AK153738.1 | - | 1 | 742 | ||
| AK154034.1 | - | 1 | 373 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM936195.1 | - | 1 | 307 | Chr. 4 - 142084418 - 142085728 [+] on Build GRCm38 | |

Tmem51os1
Genomic Map