siRNA Details
| Gene Symbol: Gm16287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033543.1 | 3 | 1379 | |||
| GenBank mRNA | AK141099.1 | - | 3 | 1380 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16287| Gene Symbol: Gm16287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033543.1 | 3 | 1379 | |||
| GenBank mRNA | AK141099.1 | - | 3 | 1380 | ||