siRNA Details
| Gene Symbol: 4933407E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045819.1 | 3 | 541 | |||
| GenBank mRNA | AK160260.1 | - | 3 | 530 | ||
| BB616969.1 | - | 3 | 530 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933407E24Rik| Gene Symbol: 4933407E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045819.1 | 3 | 541 | |||
| GenBank mRNA | AK160260.1 | - | 3 | 530 | ||
| BB616969.1 | - | 3 | 530 | |||