siRNA Details
| Gene Symbol: Lrp8os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140147.1 | - | 442 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrp8os2| Gene Symbol: Lrp8os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140147.1 | - | 442 | |||