siRNA Details
| Gene Symbol: 8030451A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033210.1 | - | 156 | |||
| AK038610.1 | - | 70 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8030451A03Rik| Gene Symbol: 8030451A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033210.1 | - | 156 | |||
| AK038610.1 | - | 70 | ||||