siRNA Details
| Gene Symbol: Gm10631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135158.1 | - | 2, 3 | 330 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10631| Gene Symbol: Gm10631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135158.1 | - | 2, 3 | 330 | ||