siRNA Details
Gene Symbol: Gm5544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142329.1 | - | 2 | 500 | ||
BC147112.1 | - | 1 | 333 | 9056126 | ||
BC147113.1 | - | 1 | 333 | 9056128 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142329.1 | - | 2 | 500 | ||
BC147112.1 | - | 1 | 333 | 9056126 | ||
BC147113.1 | - | 1 | 333 | 9056128 |