siRNA Details
| Gene Symbol: Gm15441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040409.1 | 3 | 1291 | |||
| GenBank mRNA | AK143345.1 | - | 2 | 275 | ||
| AK143784.1 | - | 3 | 1290 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15441| Gene Symbol: Gm15441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040409.1 | 3 | 1291 | |||
| GenBank mRNA | AK143345.1 | - | 2 | 275 | ||
| AK143784.1 | - | 3 | 1290 | |||