siRNA Details
| Gene Symbol: Gm15417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040403.1 | 2 | 812 | |||
| NR_040404.1 | 2 | 273 | ||||
| GenBank mRNA | AK136654.1 | - | 2 | 274 | ||
| AK161599.1 | - | 2 | 813 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15417| Gene Symbol: Gm15417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040403.1 | 2 | 812 | |||
| NR_040404.1 | 2 | 273 | ||||
| GenBank mRNA | AK136654.1 | - | 2 | 274 | ||
| AK161599.1 | - | 2 | 813 | |||